Anti-Nucleocapsid protein Hybridoma Polyclonal Antibody (AS57) Liquid

Supplier: ACROBIOSYSTEMS INC MS
NUNS57100MG NUNS5710MG
103889-250EA 172307.69 USD
103889-250 103889-752
Anti-Nucleocapsid protein Hybridoma Polyclonal Antibody (AS57) Liquid
Antibodies
Type: Primary
Antigen: Nucleocapsid protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR