Anti-S2 protein CHO Polyclonal Antibody

Supplier: ACROBIOSYSTEMS INC MS
S2NS861MG S2NS86100UG
103888-942EA 3047.69 USD
103888-942 103888-944
Anti-S2 protein CHO Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S2 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: CHO
Isotype: Human IgG4 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR