Anti-S protein RBD HEK293 Polyclonal Antibody IgG1

Supplier: ACROBIOSYSTEMS INC MS
SPDM1801MG SPDM180100UG
103888-922EA 3906 USD
103888-922 103888-924
Anti-S protein RBD HEK293 Polyclonal Antibody IgG1
Antibodies
Type: Primary
Antigen: S protein RBD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR