Transfer Pipettes, Individual Wrap, Sterile, Cole-Parmer
Supplier: Antyila Scientific
06226-42 06226-43 06226-44 06226-45
76476-420BX 69.86 USD
76476-420 76476-422 76476-424 76476-426
Transfer Pipettes, Individual Wrap, Sterile, Cole-Parmer
Pipettes Pasteur/Transfer Pipettes
No more breakage or cross-contamination.
  • Translucent shatterproof LDPE is nontoxic and biologically inert
  • Nonbinding for cells and proteins to prevent sample loss
  • Heat sealable and liquid nitrogen compatible
  • Sterilization method: ethylene oxide
Low-density polyethylene (LDPE) makes these sterile pipettes with built-in bulbs the perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glassMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable