D-Cyclopropylalinine 95%

Supplier: Ambeed
Warning

Synonyms: β-Cyclopropyl-D-Ala-OH, H-β-Cyclopropyl-D-Ala-OH

A197601-1G A197601-5G A197601-10G A197601-250MG A197601-25G
103816-950EA 125 USD
103816-950 103847-572 77085-588 77085-590 77152-052 77152-054 77302-558
D-Cyclopropylalinine 95%
D-Cyclopropylalinine

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.2 g/mol
Boiling Pt: 258 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02259520
CAS Number: 121786-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR