β-Alanine 95%

Supplier: Ambeed
Warning

A146854-100G A146854-500G A146854-1000G A146854-25G
103808-678EA 1063 USD
103808-678 76997-112 76997-114 76997-116 77299-548 77299-550
β-Alanine 95%
β-Alanine

Precaución: for Research Use Only

Formula: C₃H₇NO₂
Boiling Pt: 237.1±23.0 °C (760 mm Hg)
Melting Pt: 202 °C
Flash Pt: 97.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR