Z-Thr-OMe 97%

Supplier: Ambeed
Warning

Synonyms: Methyl ((benzyloxy)carbonyl)-L-threoninate, N-Benzyloxycarbonyl-L-threonine methyl ester

A218672-10G A218672-1G A218672-5G A218672-25G A218672-100G A218672-500G
103818-726EA 5 USD
103818-726 103849-596 76721-892 77088-122 77088-124 77303-498 77667-826 77667-828
Z-Thr-OMe 97%
Z-Thr-OMe

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₅
MW: 267.281 g/mol
Boiling Pt: 444.7 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 222.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00038585
CAS Number: 57224-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR