O-Benzyl-N-benzyloxycarbonyl-L-threonine 97%

Supplier: Ambeed
Warning

Synonyms: Z-Thr(Bzl)-OH, Cbz-Thr(Bzl)-OH, (2S,3R)-3-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)butanoic acid

A293256-250MG A293256-1G
103813-974EA 10 USD
103813-974 103835-112 76816-612 77305-624
O-Benzyl-N-benzyloxycarbonyl-L-threonine 97%
O-Benzyl-N-benzyloxycarbonyl-L-threonine

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO₅
MW: 343.379 g/mol
Boiling Pt: 539.3 °C (760 mm Hg)
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00077042
CAS Number: 69863-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR