N-Carbobenzoxy-O-tert-butyl-L-serine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-Ser(tBu)-OH, N-Cbz-O-tert-butyl-L-serine, (S)-2-(((Benzyloxy)carbonyl)amino)-3-(tert-butoxy)propanoic acid, Cbz-Ser-OtBu
A114109-100G
A114109-25G
A114109-1G
103798-350EA
5
USD
103798-350
103824-192
77025-960
77025-962
77311-014
N-Carbobenzoxy-O-tert-butyl-L-serine 97%
N-Carbobenzoxy-O-tert-butyl-L-serine
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₅ MW: 295.34 g/mol Boiling Pt: 465.3 °C (760 mm Hg) Melting Pt: 148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038275 CAS Number: 1676-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...