N-Carbobenzoxy-L-serine methyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-Ser-OMe, CBZ-L-Serine methyl ester, N-Cbz-L-serine Methyl Ester, Methyl ((benzyloxy)carbonyl)-L-serinate
A244082-100G
A244082-10G
A244082-500G
A244082-1G
A244082-5G
103811-566EA
6
USD
103811-566
103851-394
77000-512
77000-514
77000-516
77090-600
77090-602
N-Carbobenzoxy-L-serine methyl ester 97%
N-Carbobenzoxy-L-serine methyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₅ MW: 253.25 g/mol Boiling Pt: 442.6 °C (760 mm Hg) Melting Pt: 41…43 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077039 CAS Number: 1676-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...