N-Carbobenzoxy-L-serine benzyl ester 98%
Supplier: Ambeed
|
Warning
|
A133078-100G
A133078-10G
A133078-25G
A133078-5G
A133078-500G
103801-818EA
5
USD
103801-818
77027-152
77027-154
77314-414
77314-416
77670-894
N-Carbobenzoxy-L-serine benzyl ester 98%
N-Carbobenzoxy-L-serine benzyl ester
Precaución: for Research Use Only
Formula:
C₁₈H₁₉NO₅ Boiling Pt: 534.5 °C (760 mm Hg) Melting Pt: 83…86 °C Flash Pt: 277 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037824 CAS Number: 21209-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...