Z-Prolinol 98%

Supplier: Ambeed
Warning

A125121-100G A125121-10G A125121-25G A125121-1G
103800-626EA 6 USD
103800-626 76810-808 76810-810 76810-812 77312-774
Z-Prolinol 98%
Z-Prolinol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
Boiling Pt: 381.9 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02683224
CAS Number: 6216-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR