(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Z-L-2-phenylglycine, (S)-2-(((Benzyloxy)carbonyl)amino)-2-phenylacetic acid, N-Benzyloxycarbonyl-L-phenylglycine, Z-Phg-OH

A132662-100G A132662-10G A132662-500G A132662-25G
103808-450EA 281 USD
103808-450 77027-118 77027-120 77027-122 77314-340
(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid 97%
(S)-2-(Benzyloxycarbonylamino)-2-phenylacetic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₄
Flash Pt: 253.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00077033
CAS Number: 53990-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR