Cbz-L-ornithine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-Orn-OH, Nα-Benzyloxycarbonyl-L-ornithine, (S)-5-Amino-2-(((benzyloxy)carbonyl)amino)pentanoic acid, N-α-Cbz-L-Ornithine
A238017-100G
A238017-10G
A238017-250MG
A238017-1G
A238017-25G
103811-196EA
10
USD
103811-196
103850-980
77000-342
77000-344
77000-346
77090-080
77090-082
Cbz-L-ornithine 98%
Cbz-L-ornithine
Precaución: for Research Use Only
Formula:
C₁₃H₁₈N₂O₄ MW: 266.297 g/mol Boiling Pt: 489.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00038200 CAS Number: 2640-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...