Z-N-Me-Ala-OH 95%

Supplier: Ambeed
Warning

A206278-250MG A206278-1G A206278-5G A206278-100G A206278-10G
103817-700EA 13 USD
103817-700 103848-370 76785-518 77086-618 77086-620 77667-730 77872-039
Z-N-Me-Ala-OH 95%
Z-N-Me-Ala-OH
Formula: C₁₂H₁₅NO₄
Boiling Pt: 389.5 °C (760 mm Hg)
Flash Pt: 189.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153386
CAS Number: 21691-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR