N(α)-Benzyloxycarbonyl-L-lysine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH
A118492-100G
A118492-10G
A118492-25G
A118492-500G
A118492-1G
A118492-5G
103799-068EA
9
USD
103799-068
76995-620
76995-622
76995-624
76995-626
77297-608
77311-740
N(α)-Benzyloxycarbonyl-L-lysine 98%
N(α)-Benzyloxycarbonyl-L-lysine
Precaución: for Research Use Only
Formula:
C₁₄H₂₀N₂O₄ MW: 280.3 g/mol Boiling Pt: 497 °C (760 mm Hg) Melting Pt: 226…231 °C Flash Pt: 254.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038204 CAS Number: 2212-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...