Z-L-Lys(Tfa)-OH 97%

Supplier: Ambeed
Warning

A102137-100G A102137-10G A102137-25G A102137-1G A102137-5G
103796-312EA 9 USD
103796-312 103822-006 77148-262 77148-264 77148-266 77308-882 77308-884
Z-L-Lys(Tfa)-OH 97%
Z-L-Lys(Tfa)-OH

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₂O₅F₃
Boiling Pt: 566.161 °C (760 mm Hg)
Melting Pt: 88…95 °C
Flash Pt: 296.204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270524
CAS Number: 14905-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR