Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nepsilon-Boc-Nalpha-Cbz-L-lysine, Z-Lys(Boc)-OH
A210183-100G
A210183-10G
A210183-500G
A210183-1G
A210183-25G
A210183-5G
103817-984EA
14
USD
103817-984
103848-676
76999-592
76999-594
76999-596
77087-048
77087-050
77087-052
Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine 98%
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine
Precaución: for Research Use Only
Formula:
C₁₉H₂₈N₂O₆ MW: 380.44 g/mol Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062271 CAS Number: 2389-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...