Zileuton 97%

Supplier: Ambeed
Warning

Synonyms: N-(1-Benzo[b]thien-2-ylethyl)-N-hydroxyurea, 1-(1-(Benzo[b]thiophen-2-yl)ethyl)-1-hydroxyurea

A202709-100MG A202709-10MG A202709-250MG A202709-1G A202709-5G A202709-100G A202709-1MG
103817-408EA 46 USD
103817-408 103848-052 76785-402 76785-404 76785-406 77086-168 77086-170 77667-688 77667-690
Zileuton 97%
Zileuton

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂S
MW: 236.29 g/mol
Boiling Pt: 449.4 °C (760 mm Hg)
Melting Pt: 157…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00866097
CAS Number: 111406-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR