2-(Carbobenzoxyamino)-1-ethanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Cbz-amino)-1-ethanol, N-Z-Ethanolamine, Benzyl N-(2-Hydroxyethyl)carbamate, Benzyl (2-hydroxyethyl)carbamate, Z-Glycinol, N-(2-Hydroxyethyl)carbamic Acid Benzyl Ester
A102174-100G
A102174-10G
A102174-25G
A102174-5G
103822-018EA
6
USD
103822-018
76687-066
76687-068
76687-070
77308-892
2-(Carbobenzoxyamino)-1-ethanol 97%
2-(Carbobenzoxyamino)-1-ethanol
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol Melting Pt: 58…60 °C Flash Pt: 179.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191060 CAS Number: 77987-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...