5-tert-Butyl-N-carbobenzoxy-L-glutamate 97%

Supplier: Ambeed
Warning

Synonyms: Z-Glu(OtBu)-OH, N-Carbobenzoxy-L-glutamic Acid 5-tert-Butyl Ester, N-Cbz-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Cbz-L-glutamate

A117975-1000G A117975-100G A117975-10G A117975-25G A117975-500G A117975-1G A117975-5G
103798-968EA 5 USD
103798-968 103824-858 77108-374 77108-376 77108-378 77108-380 77108-382 77297-566 77311-646
5-tert-Butyl-N-carbobenzoxy-L-glutamate 97%
5-tert-Butyl-N-carbobenzoxy-L-glutamate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₆
Boiling Pt: 522.6 °C (760 mm Hg)
Melting Pt: 83…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038274
CAS Number: 3886-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR