5-Methyl-N-carbobenzoxy-L-glutamate 98%

Supplier: Ambeed
Warning

Synonyms: Z-Glu(OMe)-OH, N-Carbobenzoxy-L-glutamic Acid 5-Methyl Ester, N-Cbz-L-glutamic Acid 5-Methyl Ester, 5-Methyl N-Cbz-L-glutamate

A547649-1G A547649-5G A547649-250MG A547649-10G
103828-672EA 9 USD
103828-672 103839-794 76843-858 76843-860 76980-226 77673-458
5-Methyl-N-carbobenzoxy-L-glutamate 98%
5-Methyl-N-carbobenzoxy-L-glutamate

Precaución: For research use only

Formula: C₁₄H₁₇NO₆
MW: 295.29 g/mol
Boiling Pt: 510.6 °C (760 mm Hg)
Flash Pt: 262.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191090
CAS Number: 4652-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR