N-Carbobenzoxy-D-methionine 97%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-D-methionine, Cbz-D-Met-OH, Z-D-Met-OH, N-Cbz-D-methionine

A231791-100G A231791-10G A231791-1G A231791-25G A231791-5G
103810-790EA 6 USD
103810-790 103850-544 77000-220 77000-222 77089-434 77089-436 77089-438
N-Carbobenzoxy-D-methionine 97%
N-Carbobenzoxy-D-methionine

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄S
MW: 283.34 g/mol
Boiling Pt: 504.7 °C (760 mm Hg)
Melting Pt: 68…72 °C
Flash Pt: 259 °C
Storage Temperature: Ambient
MDL Number: MFCD00026043
CAS Number: 28862-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR