Z-D-MEAla-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-N-Me-D-Ala-OH, Z-D-MeAla-OH, N-Benzyloxycarbonyl-N-methyl-D-alanine
A149847-100G
A149847-100MG
A149847-10G
A149847-250MG
A149847-1G
A149847-25G
103804-088EA
8
USD
103804-088
103842-216
76783-318
76783-320
76783-322
76783-324
77317-728
77317-730
Z-D-MEAla-OH 97%
Z-D-MEAla-OH
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ MW: 237.255 g/mol Melting Pt: 215…219 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153387 CAS Number: 68223-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...