N-Benzyloxycarbonyl-DL-proline 98%

Supplier: Ambeed
Warning

Synonyms: Z-DL-Pro-OH, Cbz-DL-Pro-OH

A154450-10G A154450-250MG A154450-1G A154450-25G A154450-5G A154450-100G
103804-640EA 8 USD
103804-640 103842-828 76984-770 76984-772 77318-548 77318-550 77318-552 77683-295
N-Benzyloxycarbonyl-DL-proline 98%
N-Benzyloxycarbonyl-DL-proline

Precaución: For research use only.

Formula: C₁₃H₁₅NO₄
MW: 249.266 g/mol
Boiling Pt: 432.3 °C (760 mm Hg)
Flash Pt: 215.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063229
CAS Number: 5618-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR