1-Benzyl-N-carbobenzoxy-D-glutamate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl N-Cbz-D-glutamate, Z-D-Glu-OBzl, N-CBZ-D-glutamic acid alpha-benzyl ester, N-Carbobenzoxy-D-glutamic Acid 1-Benzyl Ester, N-Cbz-D-glutamic Acid 1-Benzyl Ester, Z-D-Glu-Obzl

A111126-1G A111126-5G A111126-25G
103797-796EA 5 USD
103797-796 103823-608 77284-090 77310-412 77614-858
1-Benzyl-N-carbobenzoxy-D-glutamate 98%
1-Benzyl-N-carbobenzoxy-D-glutamate

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₆
Boiling Pt: 594.3 °C (760 mm Hg)
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00069647
CAS Number: 65706-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR