Z-D-Dap(Boc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Benzyloxycarbonyl-3-(Boc-amino)-D-alanine

A137041-10G A137041-250MG A137041-25G A137041-1G A137041-5G
103802-394EA 47 USD
103802-394 77109-594 77109-596 77109-598 77298-880 77315-334
Z-D-Dap(Boc)-OH 97%
Z-D-Dap(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₆
Boiling Pt: 548.7 °C (760 mm Hg)
Flash Pt: 285.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318749
CAS Number: 62234-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR