N-Carbobenzyloxy-D-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Z-D-Ala-OH, N-Cbz-D-alanine

A134357-100G A134357-10G A134357-5G A134357-25G A134357-500G
77272-138EA 38.11 USD
77272-138 77272-140 77298-710 77314-754 77747-957
N-Carbobenzyloxy-D-alanine 98%
N-Carbobenzyloxy-D-alanine

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.2 g/mol
Boiling Pt: 422.1 °C (760 mm Hg)
Melting Pt: 83…84 °C
Flash Pt: 209.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00063126
CAS Number: 26607-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR