(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid 97%

Supplier: Ambeed
Warning

A300006-100G A300006-10G A300006-1G A300006-25G A300006-5G A300006-500G
103814-250EA 8 USD
103814-250 103835-400 76816-754 76816-756 76838-090 76838-092 76838-094 77883-822
(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid 97%
(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid
Formula: C₁₆H₂₁NO₄
Boiling Pt: 492.1 °C (760 mm Hg)
Flash Pt: 251.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00191083
CAS Number: 69901-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR