Z-Asp-OBzl 97%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, Cbz-Asp-OBzl

A189154-10G A189154-250MG A189154-1G A189154-5G A189154-25G A189154-100G
103816-164EA 7 USD
103816-164 103846-756 76974-792 76974-794 77084-514 77084-516 77302-170 77611-828
Z-Asp-OBzl 97%
Z-Asp-OBzl

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₆
Boiling Pt: 583.5 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 306.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038594
CAS Number: 4779-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR