N-Cbz-L-asparagine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-Asn-OH, Nalpha-Cbz-L-asparagine
A180583-25G
A180583-100G
A180583-10G
A180583-500G
A180583-5G
A180583-1000G
103809-144EA
249
USD
103809-144
77083-402
77274-050
77274-052
77274-054
77274-056
77301-680
N-Cbz-L-asparagine 97%
N-Cbz-L-asparagine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₂O₅ MW: 266.3 g/mol Boiling Pt: 580.6 °C (760 mm Hg) Melting Pt: 160…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008035 CAS Number: 2304-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...