Nɑ-Carbobenzoxy-L-arginine 97%

Supplier: Ambeed
Warning

A402464-25G A402464-100G A402464-10G A402464-500G
103838-112EA 6 USD
103838-112 76841-570 77278-752 77278-754 77278-756
Nɑ-Carbobenzoxy-L-arginine 97%
Nalpha-Carbobenzoxy-L-arginine

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₄O₄
Melting Pt: 171…174 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001762
CAS Number: 1234-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR