N-Carbobenzyloxy-L-alanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine
A278709-1000G
A278709-100G
A278709-10G
A278709-500G
A278709-25G
A278709-5G
103853-366EA
6
USD
103853-366
76977-350
76977-352
76977-354
76977-356
77305-302
77305-304
N-Carbobenzyloxy-L-alanine 97%
N-Carbobenzyloxy-L-alanine
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₄ Boiling Pt: 422.1 °C (760 mm Hg) Melting Pt: 82…84 °C Flash Pt: 209.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002640 CAS Number: 1142-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...