Yttrium (III) trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A129670-25G A129670-1G A129670-5G
103801-310EA 9 USD
103801-310 77298-356 77313-696 77313-698
Yttrium (III) trifluoromethanesulfonate 98%
Yttrium(III) trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₃F₉O₉S₃Y
Storage Temperature: Ambient
MDL Number: MFCD00209623
CAS Number: 52093-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR