Y-27632 dihydrochloride 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Y-27632 2HCl, (R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochloride, Rock inhibitor y-27632, trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide dihydrochloride
A101744-100MG
A101744-250MG
A101744-1G
A101744-10MG
A101744-50MG
103791-408EA
39
USD
103791-408
103791-954
103791-956
103796-110
103799-254
77160-210
77160-212
77308-706
77570-112
77610-752
Y-27632 dihydrochloride 99%
Y-27632 dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₂₁N₃O·2HCl MW: 320.26 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03490488 CAS Number: 129830-38-2 | 146986-50-7 (free base) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...