1,2,4,5-Tetraaminobenzene tetrahydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2,4,5-Tetraaminobenzene tetrahydrochloride, Benzene-1,2,4,5-tetramine 4HCl, Benzene-1,2,4,5-tetraamine tetrahydrochloride, Benzene-1,2,4,5-tetrayltetraamine tetrahydrochloride, Y15, 1,2,4,5-Benzenetetramine tetrahydrochloride, 1,2,4,5-Benzenetetramine, tetrahydrochloride
A191099-1G
A191099-5G
A191099-10MG
A191099-25G
A191099-50MG
A191099-250MG
103816-348EA
45
USD
103816-348
103846-950
77084-782
77084-784
77191-986
77191-988
77191-990
77615-624
1,2,4,5-Tetraaminobenzene tetrahydrochloride 97%
1,2,4,5-Tetraaminobenzene tetrahydrochloride
Precaución: for Research Use Only
Formula:
C₆H₂(NH₂)₄·4HCl MW: 284.006 g/mol Melting Pt: ≥300 °C Flash Pt: 233.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012970 CAS Number: 4506-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...