Valerophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1-pentanone, 1-Phenylpentan-1-one

A696222-1000G A696222-100G A696222-500G A696222-25G A696222-5G
103840-608EA 5 USD
103840-608 76791-044 76791-046 76791-048 76844-826 77308-140
Valerophenone 98%
Valerophenone

Precaución: for Research Use Only

Formula: C₆H₅C(O)(CH₂)₃CH₃
MW: 162.23 g/mol
Boiling Pt: 105…107 °C (5 mm Hg)
Melting Pt: …9 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR