Undecanoic acid 98%

Supplier: Ambeed
Warning

A320799-25G A320799-100G A320799-10G A320799-500G
103810-108EA 195 USD
103810-108 76839-064 77002-856 77002-858 77672-646
Undecanoic acid 98%
Undecanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₂
Boiling Pt: 248…250 °C (760 mm Hg)
Melting Pt: 28…31 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS Number: 112-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR