Undecane-1,11-diol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,11-Undecanediol
A417809-25G
A417809-5G
A417809-100G
A417809-10G
A417809-500G
A417809-1G
103827-346EA
5
USD
103827-346
103838-392
76841-988
76841-990
76877-056
76877-058
76877-060
77307-196
Undecane-1,11-diol 98%
Undecane-1,11-diol
Precaución: for Research Use Only
Formula:
C₁₁H₂₄O₂ MW: 188.3 g/mol Boiling Pt: 318.7 °C (760 mm Hg) Melting Pt: 60…64 °C Flash Pt: 146.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041568 CAS Number: 765-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...