Uridine-5'-diphosphate disodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate

A177143-1MG A177143-250MG A177143-1G A177143-5G A177143-10G
103807-216EA 22 USD
103807-216 103845-542 76784-472 76784-474 77082-926 77082-928 77550-136
Uridine-5'-diphosphate disodium salt 98%
Uridine-5'-diphosphate disodium salt

Precaución: for Research Use Only

Formula: C₉H₁₂N₂Na₂O₁₂P₂
MW: 448.124 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00149253
CAS Number: 27821-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR