Tris(triphenylphosphine)rhodium(I) chloride 98%

Supplier: Ambeed
Warning

Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

A318556-100MG A318556-250MG A318556-1G A318556-5G
103825-172EA 106 USD
103825-172 103836-106 77002-806 77002-808 77306-180 77672-636
Tris(triphenylphosphine)rhodium(I) chloride 98%
Tris(triphenylphosphine)rhodium(I) chloride

Precaución: for Research Use Only

Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 157
Flash Pt: 181.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00010016
CAS Number: 14694-95-2
Merck Index: 12,02226

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR