Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct, Tris(dibenylideneacetone)dipalladium-chloroform, Tris(dibenzylideneacetone)dipalladium-chloroform adduct
A480966-1G
A480966-5G
A480966-100MG
A480966-10G
A480966-250MG
103828-122EA
25
USD
103828-122
103839-196
76843-138
76843-140
76979-744
76979-746
77613-506
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 98%
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform
Precaución: For research use only
Formula:
C₅₁H₄₂O₃Pd₂·CHCl₃ MW: 1035.11 g/mol Melting Pt: 131…135 °C Flash Pt: 176.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075479 CAS Number: 52522-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...