Tris(dibenzylideneacetone)dipalladium(0) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]
A118340-10G
A118340-250MG
A118340-25G
A118340-1G
A118340-5G
A118340-100G
103799-038EA
25
USD
103799-038
76971-352
76971-354
76971-356
77311-716
77311-718
77566-166
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)
Precaución: For research use only.
Formula:
C₅₁H₄₂O₃Pd₂ MW: 915.72 g/mol Boiling Pt: 400.7 °C (760 mm Hg) Melting Pt: 152…155 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013310 CAS Number: 51364-51-3 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...