Tris(4-methoxyphenyl)phosphine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(p-methoxyphenyl)phosphine
A354955-100G
A354955-10G
A354955-1G
A354955-5G
A354955-500G
103826-138EA
5
USD
103826-138
103837-142
76788-438
76788-440
76840-270
76840-272
77668-652
Tris(4-methoxyphenyl)phosphine 97%
Tris(4-methoxyphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₂₁H₂₁O₃P MW: 352.37 g/mol Boiling Pt: 457.8 °C (760 mm Hg) Melting Pt: 131…134 °C Flash Pt: 287.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014896 CAS Number: 855-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...