Tris(4-iodophenyl)amine 97%
Supplier: Ambeed
|
Warning
|
A310285-25G
A310285-5G
A310285-10G
A310285-1G
103814-614EA
9
USD
103814-614
103835-796
76838-572
76838-574
77032-590
77306-016
Tris(4-iodophenyl)amine 97%
Tris(4-iodophenyl)amine
Precaución: for Research Use Only
Formula:
C₁₈H₁₂I₃N Boiling Pt: 559.5 °C (760 mm Hg) Melting Pt: 167 °C Flash Pt: 292.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321198 CAS Number: 4181-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...