Tris(4-fluorophenyl)phosphine 97%

Supplier: Ambeed
Warning

A118631-100G A118631-10G A118631-25G A118631-1G A118631-5G
103799-084EA 5 USD
103799-084 76781-562 76781-564 76781-566 77311-750 77311-752
Tris(4-fluorophenyl)phosphine 97%
Tris(4-fluorophenyl)phosphine

Precaución: for Research Use Only

Formula: C₁₈H₁₂F₃P
MW: 316.26 g/mol
Boiling Pt: 351.5 °C (760 mm Hg)
Melting Pt: 79…83 °C
Flash Pt: 166.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR