Tris(4-bromophenyl)amine 98%

Supplier: Ambeed
Warning

A330533-25G A330533-5G A330533-100G A330533-10G A330533-500G
103836-516EA 7 USD
103836-516 76839-496 76839-498 77032-868 77032-870 77032-872
Tris(4-bromophenyl)amine 98%
Tris(4-bromophenyl)amine

Precaución: for Research Use Only

Formula: C₁₈H₁₂Br₃N
MW: 482.01 g/mol
Boiling Pt: 514.9 °C (760 mm Hg)
Melting Pt: 141…143 °C
Flash Pt: 265.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009665
CAS Number: 4316-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR