Tris(3-methoxyphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
A233003-100G
A233003-10G
A233003-1G
A233003-25G
A233003-5G
103810-866EA
6
USD
103810-866
103850-624
76786-336
76786-338
77089-568
77089-570
77089-572
Tris(3-methoxyphenyl)phosphine 98%
Tris(3-methoxyphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₂₁H₂₁O₃P Boiling Pt: 476.3 °C (760 mm Hg) Melting Pt: 113…115 °C Flash Pt: 301.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008386 CAS Number: 29949-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...