Tris(2-pyridylmethyl)amine 97%

Supplier: Ambeed
Warning

A307393-1G A307393-5G A307393-100G A307393-100MG A307393-10G A307393-250MG A307393-25G
103814-502EA 10 USD
103814-502 103835-678 76838-420 76838-422 77154-292 77154-294 77154-296 77154-298 77305-956
Tris(2-pyridylmethyl)amine 97%
Tris(2-pyridylmethyl)amine

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₄
MW: 290.37 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14708176
CAS Number: 16858-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR