Tris(2-methoxyphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(o-methoxyphenyl)phosphine
A159874-100G
A159874-10G
A159874-1G
A159874-25G
A159874-5G
A159874-250MG
103805-298EA
6
USD
103805-298
103843-524
76783-786
76783-788
77319-426
77319-428
77319-430
77592-946
Tris(2-methoxyphenyl)phosphine 98%
Tris(2-methoxyphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₂₁H₂₁O₃P Boiling Pt: 477.3 °C (760 mm Hg) Melting Pt: 198…204 °C Flash Pt: 302.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014892 CAS Number: 4731-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...